missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexyloxyaniline, 99%, Thermo Scientific Chemicals

Catalog Number 15482248
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
39905-57-2
C12H19NO
193.29
MFCD00007868
DJRKHTCUXRGYEU-UHFFFAOYSA-N
4-hexyloxyaniline, 4-hexyloxy aniline, p-hexyloxyaniline, benzenamine, 4-hexyloxy, 4-n-hexyloxyaniline, p-hexyloxy aniline, 4-hexyloxy benzenamine, unii-vmx449yp3l, aniline, p-hexyloxy, vmx449yp3l
38360
4-hexoxyaniline
CCCCCCOC1=CC=C(N)C=C1
This item is not returnable. View return policy
39905-57-2
C12H19NO
193.29
MFCD00007868
DJRKHTCUXRGYEU-UHFFFAOYSA-N
4-hexyloxyaniline, 4-hexyloxy aniline, p-hexyloxyaniline, benzenamine, 4-hexyloxy, 4-n-hexyloxyaniline, p-hexyloxy aniline, 4-hexyloxy benzenamine, unii-vmx449yp3l, aniline, p-hexyloxy, vmx449yp3l
38360
4-hexoxyaniline
CCCCCCOC1=CC=C(N)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 43°C to 45°C
Color White to Brown
Boiling Point 155°C to 158°C (5 mmHg)
Flash Point 113°C (235°F)
Quantity 1 g
Formula Weight 193.29
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 4-n-Hexyloxyaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'