missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexylbenzeneboronic acid, 97%, Thermo Scientific™

Catalog Number p-7055865
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
105365-50-2
C12H19BO2
206.092
MFCD04039029
CFYBKGAFWFRFRJ-UHFFFAOYSA-N
4-hexylphenyl boronic acid, 4-n-hexylphenylboronic acid, 4-hex-1-yl benzeneboronic acid, 1-borono-4-hex-1-yl benzene, 4-n-hexylbenzeneboronic acid, boronic acid, 4-hexylphenyl, 4-hexylphenylboronicacid, p-hexylphenylboronic acid, 4-hexylbenzeneboronic acid, 4-hexylphenyl-boronic acid
4197864
(4-hexylphenyl)boronic acid
B(C1=CC=C(C=C1)CCCCCC)(O)O
This item is not returnable. View return policy
105365-50-2
C12H19BO2
206.092
MFCD04039029
CFYBKGAFWFRFRJ-UHFFFAOYSA-N
4-hexylphenyl boronic acid, 4-n-hexylphenylboronic acid, 4-hex-1-yl benzeneboronic acid, 1-borono-4-hex-1-yl benzene, 4-n-hexylbenzeneboronic acid, boronic acid, 4-hexylphenyl, 4-hexylphenylboronicacid, p-hexylphenylboronic acid, 4-hexylbenzeneboronic acid, 4-hexylphenyl-boronic acid
4197864
(4-hexylphenyl)boronic acid
B(C1=CC=C(C=C1)CCCCCC)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 206.09
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-n-Hexylbenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'