missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-n-Heptylbenzoyl chloride, 97%

Brand:  Alfa Aesar™ A11942.14

Code : NEW

Additional Details : CAS Number : 50606-96-7 Weight : 0.02500kg Transport : UN number : 3265 Chem class : 8 Pack group : II

Quantity 25g

 View more versions of this product

Product Code. 11483934

  • £125.93 / 25g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

4-n-Heptylbenzoyl chloride
C14H19ClO
MFCD00000702
2092107
50606-96-7
238.76
UN3265

Specifications

4-n-Heptylbenzoyl chloride
196°C (40mmHg)
50606-96-7
C14H19ClO
MFCD00000702
2092107
>110°C (230°F)
25g
97%
238.76
UN3265
Videos
SDS