missing translation for 'onlineSavingsMsg'
Learn More

4-(n-Butylsulfamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog Number p-7035501
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
850589-32-1
C10H16BNO4S
257.111
MFCD07363753
BEBIGVQNUYZEPK-UHFFFAOYSA-N
n-butyl 4-boronobenzenesulfonamide, 4-n-butylsulfamoyl phenyl boronic acid, 4-n-butylsulphonamido benzeneboronic acid, 4-butylsulfamoyl phenylboronic acid, acmc-209q2v, 4-butylsulfamoyl phenyl boronic acid, n-n-butyl 4-boronobenzenesulfonamide, 4-n-butylsulfamoyl phenylboronic acid, 4-n-n-butylsulfamoylphenylboronic acid, 4-n-butylsulfamoyl benzeneboronic acid
44119412
[4-(butylsulfamoyl)phenyl]boronic acid
B(C1=CC=C(C=C1)S(=O)(=O)NCCCC)(O)O
This item is not returnable. View return policy
850589-32-1
C10H16BNO4S
257.111
MFCD07363753
BEBIGVQNUYZEPK-UHFFFAOYSA-N
n-butyl 4-boronobenzenesulfonamide, 4-n-butylsulfamoyl phenyl boronic acid, 4-n-butylsulphonamido benzeneboronic acid, 4-butylsulfamoyl phenylboronic acid, acmc-209q2v, 4-butylsulfamoyl phenyl boronic acid, n-n-butyl 4-boronobenzenesulfonamide, 4-n-butylsulfamoyl phenylboronic acid, 4-n-n-butylsulfamoylphenylboronic acid, 4-n-butylsulfamoyl benzeneboronic acid
44119412
[4-(butylsulfamoyl)phenyl]boronic acid
B(C1=CC=C(C=C1)S(=O)(=O)NCCCC)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 124°C to 126°C
Quantity 250 mg
Formula Weight 257.11
Percent Purity 97%
Chemical Name or Material 4-(n-Butylsulfamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'