missing translation for 'onlineSavingsMsg'
Learn More

4-(n-Butoxycarbonyl)phenyl isocyanate, 99%, Thermo Scientific™

Catalog Number p-7045410
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
102561-47-7
C12H13NO3
219.24
MFCD00037043
PVCGQSQNUXNTNA-UHFFFAOYSA-N
4-n-butoxycarbonyl phenyl isocyanate, acmc-20e5xd, 4-carbobutoxy phenyl isocyanate, butyl 4-isocyanatobenzoate, 4-n-butoxycarbonyl phenylisocyanate, 4-carbobutoxyphenyl isocyanate, 4-n-carbobutoxyphenyl isocyanate, benzoic acid,4-isocyanato-, butyl ester, benzoic acid, 4-isocyanato-, butyl ester 9ci, 4-n-butoxycarbonyl phenyl isocyanate; 4-carbobutoxy phenyl isocyanate
4112112
butyl 4-isocyanatobenzoate
CCCCOC(=O)C1=CC=C(C=C1)N=C=O
This item is not returnable. View return policy
102561-47-7
C12H13NO3
219.24
MFCD00037043
PVCGQSQNUXNTNA-UHFFFAOYSA-N
4-n-butoxycarbonyl phenyl isocyanate, acmc-20e5xd, 4-carbobutoxy phenyl isocyanate, butyl 4-isocyanatobenzoate, 4-n-butoxycarbonyl phenylisocyanate, 4-carbobutoxyphenyl isocyanate, 4-n-carbobutoxyphenyl isocyanate, benzoic acid,4-isocyanato-, butyl ester, benzoic acid, 4-isocyanato-, butyl ester 9ci, 4-n-butoxycarbonyl phenyl isocyanate; 4-carbobutoxy phenyl isocyanate
4112112
butyl 4-isocyanatobenzoate
CCCCOC(=O)C1=CC=C(C=C1)N=C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.105
Boiling Point 295°C
Flash Point >110°C (230°F)
Refractive Index 1.5275
Quantity 1 g
UN Number UN2206
Beilstein 7640773
Sensitivity Moisture sensitive
Formula Weight 219.24
Percent Purity 99%
Chemical Name or Material 4-(n-Butoxycarbonyl)phenyl isocyanate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'