missing translation for 'onlineSavingsMsg'
Learn More

4-N-BOC-Amino-4-carboxytetrahydropyran, 95%, Thermo Scientific Chemicals

Catalog Number p-70572
90.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


missing translation for 'orderingAttributeHoverText'
Quantity:
500 mg
2 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
172843-97-9
C11H19NO5
245.27
MFCD02683136
SPPDKPRJPFTBEV-UHFFFAOYSA-N
4-boc-amino tetrahydropyran-4-carboxylic acid, 4-n-boc-amino-4-carboxytetrahydropyran, 4-tert-butoxycarbonyl amino tetrahydro-2h-pyran-4-carboxylic acid, 4-tert-butoxycarbonyl amino oxane-4-carboxylic acid, 4-tert-butoxy carbonyl amino oxane-4-carboxylic acid, 4-tert-butoxycarbamoyl tetrahydropyran-4-carboxylic acid, 4-tert-butoxycarbonylamino tetrahydro-2h-pyran-4-carboxylic acid, 4-tert-butoxy carbonylamino-2h-3,4,5,6-tetrahydropyran-4-carboxylic acid
1268219
4-[(2-methylpropan-2-yl)oxycarbonylamino]oxane-4-carboxylic acid
CC(C)(C)OC(=O)NC1(CCOCC1)C(=O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
172843-97-9
C11H19NO5
245.27
MFCD02683136
SPPDKPRJPFTBEV-UHFFFAOYSA-N
4-boc-amino tetrahydropyran-4-carboxylic acid, 4-n-boc-amino-4-carboxytetrahydropyran, 4-tert-butoxycarbonyl amino tetrahydro-2h-pyran-4-carboxylic acid, 4-tert-butoxycarbonyl amino oxane-4-carboxylic acid, 4-tert-butoxy carbonyl amino oxane-4-carboxylic acid, 4-tert-butoxycarbamoyl tetrahydropyran-4-carboxylic acid, 4-tert-butoxycarbonylamino tetrahydro-2h-pyran-4-carboxylic acid, 4-tert-butoxy carbonylamino-2h-3,4,5,6-tetrahydropyran-4-carboxylic acid
1268219
4-[(2-methylpropan-2-yl)oxycarbonylamino]oxane-4-carboxylic acid
CC(C)(C)OC(=O)NC1(CCOCC1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 500 mg
Formula Weight 245.27
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-N-BOC-Amino-4-carboxytetrahydropyran, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4-N-BOC-Amino-4-carboxytetrahydropyran
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'