missing translation for 'onlineSavingsMsg'
Learn More

4-(Morpholinomethyl)benzaldehyde, 97%, Thermo Scientific™

Catalog Number p-3075572
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
82413-63-6
C12H15NO2
205.257
MFCD05865107
KMAHWHPUXGNVBN-UHFFFAOYSA-N
4-morpholinomethyl benzaldehyde, 4-morpholin-4-ylmethyl benzaldehyde, 4-morpholinomethyl-benzaldehyde, 4-4-morpholinylmethyl benzaldehyde, 4-morpholin-4-ylmethyl-benzaldehyde, 4-4-carboxaldehydebenzyl morpholine, benzaldehyde,4-4-morpholinylmethyl
2795498
4-(morpholin-4-ylmethyl)benzaldehyde
C1COCCN1CC2=CC=C(C=C2)C=O
This item is not returnable. View return policy
82413-63-6
C12H15NO2
205.257
MFCD05865107
KMAHWHPUXGNVBN-UHFFFAOYSA-N
4-morpholinomethyl benzaldehyde, 4-morpholin-4-ylmethyl benzaldehyde, 4-morpholinomethyl-benzaldehyde, 4-4-morpholinylmethyl benzaldehyde, 4-morpholin-4-ylmethyl-benzaldehyde, 4-4-carboxaldehydebenzyl morpholine, benzaldehyde,4-4-morpholinylmethyl
2795498
4-(morpholin-4-ylmethyl)benzaldehyde
C1COCCN1CC2=CC=C(C=C2)C=O
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 205.26
Percent Purity 95%
Chemical Name or Material 4-(Morpholinomethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'