missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinobenzenecarbothioamide, 97%, Maybridge™

£54.00 - £144.00

Chemical Identifiers

CAS 519056-60-1
Molecular Formula C11H14N2OS
Molecular Weight (g/mol) 222.31
MDL Number MFCD04115379
InChI Key KOPFTYFPHHZQCH-UHFFFAOYSA-N
Synonym 4-morpholinobenzenecarbothioamide, 4-morpholinobenzothioamide, 4-morpholin-4-yl benzenecarbothioamide, 4-morpholin-4'-y lthiobenzamide, amino 4-morpholin-4-ylphenyl methane-1-thione
PubChem CID 2795360
IUPAC Name 4-morpholin-4-ylbenzenecarbothioamide
SMILES NC(=S)C1=CC=C(C=C1)N1CCOCC1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10684935
SDS View Documents
Maybridge
CC17418CB
250mg
£54.00
250mg
View alternative products
Save Up to 
»
Estimated Shipment: 20-10-2021
Please to purchase this item. Need a web account? Register with us today!
10451274
SDS View Documents
Maybridge
CC17418DA
1g
£102.00
1g
View alternative products
Save Up to 
»
Estimated Shipment: 20-10-2021
Please to purchase this item. Need a web account? Register with us today!
10694935
SDS View Documents
Maybridge
CC17418DE
5g
£144.00
5g
View alternative products
Save Up to 
»
Estimated Shipment: 20-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

519056-60-1
222.31
KOPFTYFPHHZQCH-UHFFFAOYSA-N
2795360
NC(=S)C1=CC=C(C=C1)N1CCOCC1
C11H14N2OS
MFCD04115379
4-morpholinobenzenecarbothioamide, 4-morpholinobenzothioamide, 4-morpholin-4-yl benzenecarbothioamide, 4-morpholin-4'-y lthiobenzamide, amino 4-morpholin-4-ylphenyl methane-1-thione
4-morpholin-4-ylbenzenecarbothioamide

Specifications

4-Morpholinobenzenecarbothioamide
250mg
96.5
97%
MFCD04115379
KOPFTYFPHHZQCH-UHFFFAOYSA-N
4-morpholin-4-ylbenzenecarbothioamide
2795360
97%
Amber glass bottle
519056-60-1
100.0
C11H14N2OS
4-morpholinobenzenecarbothioamide, 4-morpholinobenzothioamide, 4-morpholin-4-yl benzenecarbothioamide, 4-morpholin-4'-y lthiobenzamide, amino 4-morpholin-4-ylphenyl methane-1-thione
NC(=S)C1=CC=C(C=C1)N1CCOCC1
222.31
222.31
SDS
Product Certifications