missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinobenzenecarbothioamide, 97%, Thermo Scientific™

Catalog Number 10684935
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
519056-60-1
C11H14N2OS
222.31
MFCD04115379
KOPFTYFPHHZQCH-UHFFFAOYSA-N
4-morpholinobenzenecarbothioamide, 4-morpholinobenzothioamide, 4-morpholin-4-yl benzenecarbothioamide, 4-morpholin-4'-y lthiobenzamide, amino 4-morpholin-4-ylphenyl methane-1-thione
2795360
4-morpholin-4-ylbenzenecarbothioamide
NC(=S)C1=CC=C(C=C1)N1CCOCC1
This item is not returnable. View return policy
519056-60-1
C11H14N2OS
222.31
MFCD04115379
KOPFTYFPHHZQCH-UHFFFAOYSA-N
4-morpholinobenzenecarbothioamide, 4-morpholinobenzothioamide, 4-morpholin-4-yl benzenecarbothioamide, 4-morpholin-4'-y lthiobenzamide, amino 4-morpholin-4-ylphenyl methane-1-thione
2795360
4-morpholin-4-ylbenzenecarbothioamide
NC(=S)C1=CC=C(C=C1)N1CCOCC1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 222.31
Percent Purity 97%
Chemical Name or Material 4-Morpholinobenzenecarbothioamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'