missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl-α-D-galactopyranoside hydrate, 99%, Thermo Scientific Chemicals

Catalog Number p-169073
Click to view available options
Quantity:
50 mg
250 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
38597-12-5
C16H18O8
338.31
MFCD03791284
YUDPTGPSBJVHCN-HWSQZMMLNA-N
4-methylumbelliferyl-alpha-d-galactopyranoside, 4-methyl-7-2r,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy-2h-chromen-2-one, 4-methyl-7-2r,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one, 7-alpha-d-galactopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl a-d-galactopyranoside, 4-methylumbelliferyl alpha-d-galactoside, 4-methylumbelliferyl-a-d-galactopyranoside, 4-methylumbelliferyl alpha-d-galactopyranoside, 4-methyl-2-oxo-2h-1-benzopyran-7-yl alpha-d-galactopyranoside, 4-methylumbelliferyl alpha-d-galactopyranoside tlc
2733788
4-methyl-7-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
CC1=CC(=O)OC2=CC(O[C@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12
This item is not returnable. View return policy
38597-12-5
C16H18O8
338.31
MFCD03791284
YUDPTGPSBJVHCN-HWSQZMMLNA-N
4-methylumbelliferyl-alpha-d-galactopyranoside, 4-methyl-7-2r,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy-2h-chromen-2-one, 4-methyl-7-2r,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one, 7-alpha-d-galactopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl a-d-galactopyranoside, 4-methylumbelliferyl alpha-d-galactoside, 4-methylumbelliferyl-a-d-galactopyranoside, 4-methylumbelliferyl alpha-d-galactopyranoside, 4-methyl-2-oxo-2h-1-benzopyran-7-yl alpha-d-galactopyranoside, 4-methylumbelliferyl alpha-d-galactopyranoside tlc
2733788
4-methyl-7-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
CC1=CC(=O)OC2=CC(O[C@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 50 mg
Formula Weight 338.31
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylumbelliferyl-α-D-galactopyranoside hydrate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'