missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl acetate, 99%, Thermo Scientific™

Catalog Number p-7081949
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2747-05-9
This item is not returnable. View return policy
2747-05-9

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 147°C to 152°C
Molecular Formula C10H22ClNO2
MDL Number MFCD00038906
Quantity 5 g
Beilstein 189667
Synonym h-leu-otbu.hcl, l-leucine tert-butyl ester hydrochloride, h-leu-otbu hcl, l-leucine t-butyl ether hcl salt, l-leucine t-butyl ester hydrochloride, l-leucine tert-butyl ester hcl, tert-butyl l-leucinate hydrochloride, s-tert-butyl 2-amino-4-methylpentanoate hydrochloride, tert-butyl 2s-2-amino-4-methylpentanoate hydrochloride, h-leu-obut hcl
InChI Key RFUWRXIYTQGFGA-QRPNPIFTSA-N
SMILES Cl.CC(C)C[C@H](N)C(=O)OC(C)(C)C
IUPAC Name tert-butyl (2S)-2-amino-4-methylpentanoate;hydrochloride
Molecular Weight (g/mol) 223.74
PubChem CID 12451443
Formula Weight 218.21
Percent Purity 99%
Chemical Name or Material 4-Methylumbelliferyl acetate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'