missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylumbelliferone, 97%, Thermo Scientific Chemicals
17.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
4-Methylumbelliferone is used in the determination of nitric acid and as a standard for fluorometric determination of enzyme activity. It is also used in bile therapy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methylumbelliferone is used in the determination of nitric acid and as a standard for fluorometric determination of enzyme activity. It is also used in bile therapy.
Solubility
Soluble in alcohol. Insoluble in cold water. Soluble in acetic acid, alkali and ammonium hydroxide. Slightly soluble in hot water, chloroform and diethyl ether.
Notes
Store in a cool place. Incompatible with strong bases and strong oxidizing agents.
4-Methylumbelliferone is used in the determination of nitric acid and as a standard for fluorometric determination of enzyme activity. It is also used in bile therapy.
Solubility
Soluble in alcohol. Insoluble in cold water. Soluble in acetic acid, alkali and ammonium hydroxide. Slightly soluble in hot water, chloroform and diethyl ether.
Notes
Store in a cool place. Incompatible with strong bases and strong oxidizing agents.
Specifications
Melting Point | 186°C to 191°C |
Quantity | 50 g |
Beilstein | 142217 |
Merck Index | 14,4854 |
Solubility Information | Soluble in alcohol; Insoluble in cold water; Soluble in acetic acid,alkali and ammonium hydroxide; Slightly soluble in hot water,chloroform and diethyl ether. |
Formula Weight | 176.17 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methylumbelliferone |
Safety and Handling
Product Identifier
- 4-Methylumbelliferone
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : 7-hydroxy-4-methyl-1-Benzopyran-2-one
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'