missing translation for 'onlineSavingsMsg'
Learn More

4-Methylthiophene-2-carboxylic acid, 97%, Thermo Scientific™

Catalog Number 10292782
Click to view available options
Amber Glass Bottle; 250mg
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 14282-78-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H6O2S
MDL Number MFCD00089284
Quantity 250mg
Synonym 4-methyl-2-thiophenecarboxylic acid, 4-methyl-thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 4-methyl, 2-carboxy-4-methylthiophene, pubchem9431, 4-methyl-2-thenoic acid, 4-methyl-2-thiophenecarboxylicacid, 4-methyl-2-thiopene carboxylic acid, 2-thiophenecarboxylicacid, 4-methyl, 4-methyl 2-thiophene carboxylic acid
InChI Key YCIVJGQMXZZPAB-UHFFFAOYSA-N
SMILES CC1=CSC(=C1)C(=O)O
IUPAC Name 4-methylthiophene-2-carboxylic acid
Molecular Weight (g/mol) 142.172
PubChem CID 2794584
Formula Weight 142.18
Percent Purity 97%
Chemical Name or Material 4-Methylthiophene-2-carboxylic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'