missing translation for 'onlineSavingsMsg'
Learn More

4-Methylthiophene-2-carboxylic acid, 97%, Maybridge

Brand:  Maybridge GK01984DA

Code : 31

Additional Details : CAS Number : 14282-78-1 Weight : 0.05100kg

Quantity 1g

 View more versions of this product

Product Code. 10522652

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

4-Methylthiophene-2-carboxylic acid
96.5
97%
C6H6O2S
MFCD00089284
14282-78-1
100.0
97%
142.18

Specifications

4-Methylthiophene-2-carboxylic acid
14282-78-1
100.0
97%
142.18
1g
96.5
97%
C6H6O2S
MFCD00089284
Videos
SDS