missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Methylthio)benzyl chloride, 99%

Brand:  Alfa Aesar™ L19877.06

Code : NEW

Additional Details : CAS Number : 874-87-3 Weight : 0.00500kg Transport : UN number : 3265 Chem class : 8 Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 11309895

  • £97.90 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Description

Description

Applications
4-(Methylthio)benzyl chloride is used as pharmaceutical intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
Specifications

Chemical Identifiers

4-(Methylthio)benzyl chloride
C8H9ClS
MFCD00270118
4-(Chloromethyl)thioanisole
874-87-3
172.68
UN3265

Specifications

4-(Methylthio)benzyl chloride
99%
172.68
UN3265
132°C (22mmHg)
>110°C (230°F)
874-87-3
C8H9ClS
MFCD00270118
4-(Chloromethyl)thioanisole
5g
Videos
SDS