missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-168382
12.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3446-89-7
C8H8OS
152.21
MFCD00006948
QRVYABWJVXXOTN-UHFFFAOYSA-N
4-methylthio benzaldehyde, benzaldehyde, 4-methylthio, 4-methylsulfanyl benzaldehyde, 4-methylmercaptobenzaldehyde, p-methylthio benzaldehyde, p-methylthiobenzaldehyde, unii-1tvq11biy8, 4-aldehyde thioanisole, p-methylmercaptobenzaldehyde
76985
4-methylsulfanylbenzaldehyde
CSC1=CC=C(C=C1)C=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3446-89-7
C8H8OS
152.21
MFCD00006948
QRVYABWJVXXOTN-UHFFFAOYSA-N
4-methylthio benzaldehyde, benzaldehyde, 4-methylthio, 4-methylsulfanyl benzaldehyde, 4-methylmercaptobenzaldehyde, p-methylthio benzaldehyde, p-methylthiobenzaldehyde, unii-1tvq11biy8, 4-aldehyde thioanisole, p-methylmercaptobenzaldehyde
76985
4-methylsulfanylbenzaldehyde
CSC1=CC=C(C=C1)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 6.0°C
Color Yellow
Density 1.1400g/mL
Boiling Point 86.0°C to 90.0°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3SC6H4CHO
Refractive Index 1.6452
Quantity 5 mL
Beilstein 08, I, 533
Specific Gravity 1.14
Formula Weight 152.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(Methylthio)benzaldehyde, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'