Learn More
Alfa Aesar™ 4-Methylthiazole-2-carboxylic acid, 98%
Brand: Alfa Aesar™ H63065.03
Code : NEW
Additional Details : CAS Number : 14542-16-6 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
4-Methylthiazole-2-carboxylic acid | |
14542-16-6 | |
143.16 | |
MFCD07377486 |
Specifications
4-Methylthiazole-2-carboxylic acid | |
1g | |
Solid | |
143.16 |
90° to 92°C | |
14542-16-6 | |
98% | |
MFCD07377486 |