missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methylthiazole-2-carboxylic acid, 98%

Brand:  Alfa Aesar™ H63065.06

Code : NEW

Additional Details : CAS Number : 14542-16-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15419548

  • £199.28 / 5g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Methylthiazole-2-carboxylic acid
14542-16-6
143.16
MFCD07377486

Specifications

4-Methylthiazole-2-carboxylic acid
5g
Solid
143.16
90° to 92°C
14542-16-6
98%
MFCD07377486
Videos
SDS