missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)phenol, 95%, Thermo Scientific™

Catalog Number p-3736996
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
14763-60-1
C7H8O3S
172.20
MFCD00047806
KECCFSZFXLAGJS-UHFFFAOYSA-N
4-methylsulfonyl phenol, 4-methanesulfonylphenol, 4-methanesulfonyl-phenol, 4-hydroxyphenyl methyl sulfone, 4-methylsulphonylphenol, phenol, p-methylsulfonyl, phenol, 4-methylsulfonyl, p-hydroxyphenyl methyl sulfone, p-methylsulfonyl phenol, unii-f7bif7zqj9
123350
4-methylsulfonylphenol
CS(=O)(=O)C1=CC=C(O)C=C1
This item is not returnable. View return policy
14763-60-1
C7H8O3S
172.20
MFCD00047806
KECCFSZFXLAGJS-UHFFFAOYSA-N
4-methylsulfonyl phenol, 4-methanesulfonylphenol, 4-methanesulfonyl-phenol, 4-hydroxyphenyl methyl sulfone, 4-methylsulphonylphenol, phenol, p-methylsulfonyl, phenol, 4-methylsulfonyl, p-hydroxyphenyl methyl sulfone, p-methylsulfonyl phenol, unii-f7bif7zqj9
123350
4-methylsulfonylphenol
CS(=O)(=O)C1=CC=C(O)C=C1
CAS Min % 94.0
CAS Max % 100.0
Melting Point 90.0°C to 95.0°C
Color Brown-Pink to Tan
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H4SO2CH3
Quantity 2.5 g
Solubility Information Solubility in water: slight. Other solubilities: soluble in dichloromethane,methanol
Formula Weight 172.2
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-(Methylsulfonyl)phenol
Show More Show Less
Product Identifier
  • 4-(Methylsulfonyl)phenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'