missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsulphonylbenzyl bromide, Tech., Thermo Scientific Chemicals

Catalog Number p-3736994
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
53606-06-7
C8H9BrO2S
249.13
MFCD00185849
HGKPAXHJTMHWAH-UHFFFAOYSA-N
1-bromomethyl-4-methylsulfonyl benzene, 4-methylsulfonyl benzyl bromide, 1-bromomethyl-4-methanesulfonylbenzene, benzene, 1-bromomethyl-4-methylsulfonyl, 4-methylsulphonylbenzyl bromide, 4-methylsulfonyl benzylbromide, 1-bromomethyl-4-methanesulfonyl-benzene, 1-bromomethyl-4-methylsulfonylbenzene, pubchem18666, acmc-1b0ix
2733581
1-(bromomethyl)-4-methylsulfonylbenzene
CS(=O)(=O)C1=CC=C(C=C1)CBr
This item is not returnable. View return policy
53606-06-7
C8H9BrO2S
249.13
MFCD00185849
HGKPAXHJTMHWAH-UHFFFAOYSA-N
1-bromomethyl-4-methylsulfonyl benzene, 4-methylsulfonyl benzyl bromide, 1-bromomethyl-4-methanesulfonylbenzene, benzene, 1-bromomethyl-4-methylsulfonyl, 4-methylsulphonylbenzyl bromide, 4-methylsulfonyl benzylbromide, 1-bromomethyl-4-methanesulfonyl-benzene, 1-bromomethyl-4-methylsulfonylbenzene, pubchem18666, acmc-1b0ix
2733581
1-(bromomethyl)-4-methylsulfonylbenzene
CS(=O)(=O)C1=CC=C(C=C1)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 82°C to 86°C
Color Beige to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 2.5 g
Beilstein 06, IV, 2210
Formula Weight 249.13
Percent Purity ≥60%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylsulphonylbenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylsulphonylbenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'