missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)phenylhydrazine Hydrochloride, 95%, Thermo Scientific Chemicals

Catalog Number p-3736998
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
17852-67-4
C7H10N2O2S·HCl
222.69
MFCD00216494
QVCMFSJTNVQJFG-UHFFFAOYSA-N
4-methylsulfonyl phenyl hydrazine hydrochloride, 4-methylsulfonyl phenylhydrazine hydrochloride, 4-methylsulphonyl phenylhydrazine hydrochloride, 4-methylsulphonylphenylhydrazine hydrochloride, 4-methanesulfonylphenyl hydrazine hydrochloride, 4-methylsulfonylphenylhydrazine hydrochloride, hydrazine, 4-methylsulfonyl phenyl-, monohydrochloride, 4-methylsulfonyl phenylhydrazine, chloride, 4-methylsulfonyl phenylhydrazine hydrochloric acid
2735181
(4-methylsulfonylphenyl)hydrazine;hydrochloride
CS(=O)(=O)C1=CC=C(C=C1)NN.Cl
This item is not returnable. View return policy
17852-67-4
C7H10N2O2S·HCl
222.69
MFCD00216494
QVCMFSJTNVQJFG-UHFFFAOYSA-N
4-methylsulfonyl phenyl hydrazine hydrochloride, 4-methylsulfonyl phenylhydrazine hydrochloride, 4-methylsulphonyl phenylhydrazine hydrochloride, 4-methylsulphonylphenylhydrazine hydrochloride, 4-methanesulfonylphenyl hydrazine hydrochloride, 4-methylsulfonylphenylhydrazine hydrochloride, hydrazine, 4-methylsulfonyl phenyl-, monohydrochloride, 4-methylsulfonyl phenylhydrazine, chloride, 4-methylsulfonyl phenylhydrazine hydrochloric acid
2735181
(4-methylsulfonylphenyl)hydrazine;hydrochloride
CS(=O)(=O)C1=CC=C(C=C1)NN.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 202°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NHNC6H4SO2CH3·HCl
Quantity 2.5 g
Solubility Information Solubility in water: soluble
Formula Weight 222.69
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylsulfonyl)phenylhydrazine hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'