missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsalicylic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-2690723
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
50-85-1
MFCD00002454
NJESAXZANHETJV-UHFFFAOYSA-N
4-methylsalicylic acid, m-cresotic acid, 2,4-cresotic acid, m-cresotinic acid, 2-hydroxy-p-toluic acid, m-homosalicylic acid, 4-methyl-2-hydroxybenzoic acid, gamma-cresotic acid, benzoic acid, 2-hydroxy-4-methyl, 4-methylsalicylate
5788
CHEBI:20450
2-hydroxy-4-methylbenzoic acid
CC1=CC(=C(C=C1)C(=O)O)O
This item is not returnable. View return policy
50-85-1
MFCD00002454
NJESAXZANHETJV-UHFFFAOYSA-N
4-methylsalicylic acid, m-cresotic acid, 2,4-cresotic acid, m-cresotinic acid, 2-hydroxy-p-toluic acid, m-homosalicylic acid, 4-methyl-2-hydroxybenzoic acid, gamma-cresotic acid, benzoic acid, 2-hydroxy-4-methyl, 4-methylsalicylate
5788
CHEBI:20450
2-hydroxy-4-methylbenzoic acid
CC1=CC(=C(C=C1)C(=O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 175.0°C to 179.0°C
Color Beige-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H8O3
Linear Formula CH3C6H3(OH)CO2H
Quantity 25 g
Beilstein 10, 233
Merck Index 15, 2569
Molecular Weight (g/mol) 152.15
Formula Weight 152.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Methylsalicylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylsalicylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'