missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylpyrimidine, 97%, Thermo Scientific™
Click to view available options
10g; Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS | 3438-46-8 |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Density | 1.0310g/mL |
Boiling Point | 141°C to 142°C |
Flash Point | 40°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Molecular Formula | C5H6N2 |
Show More |
Safety and Handling
Product Identifier
- 4-Methylpyrimidine
- Danger
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- H360D-May damage the unborn child.
- H226-Flammable liquid and vapour.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'