missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methylpiperazine-1-carbonyl chloride, 97%

Brand:  Alfa Aesar™ H55732.14

Code : NEW

Additional Details : CAS Number : 39539-66-7 Weight : 0.02500kg Transport : UN number : 3265 Chem class : 1a Pack group : II

Quantity 25g

 View more versions of this product

Product Code. 15402068

  • £373.54 / 25g


Estimated Shipment date
from Supplier 14-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Methylpiperazine-1-carbonyl chloride
162.62
UN3265
39539-66-7
MFCD01320509

Specifications

4-Methylpiperazine-1-carbonyl chloride
25g
1.200g/mL
Liquid
162.62
UN3265
278° to 279°C
Air Sensitive
39539-66-7
1.4980
97%
MFCD01320509
Colorless to brown
Videos
SDS