missing translation for 'onlineSavingsMsg'
Learn More

(4-Methylphenoxy)acetic acid, 99%, Thermo Scientific™

Catalog Number p-3211022
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
940-64-7
C9H10O3
166.18
MFCD00014365
SFTDDFBJWUWKMN-UHFFFAOYSA-N
4-methylphenoxy acetic acid, 4-methylphenoxyacetic acid, 2-4-methylphenoxy acetic acid, p-methylphenoxyacetic acid, unii-wwy23322ik, acetic acid, 4-methylphenoxy, 2-p-tolyloxy acetic acid, p-toloxyacetic acid, acmc-20apmz, p-tolyloxy-acetic acid
70329
2-(4-methylphenoxy)acetic acid
CC1=CC=C(C=C1)OCC(=O)O
This item is not returnable. View return policy
940-64-7
C9H10O3
166.18
MFCD00014365
SFTDDFBJWUWKMN-UHFFFAOYSA-N
4-methylphenoxy acetic acid, 4-methylphenoxyacetic acid, 2-4-methylphenoxy acetic acid, p-methylphenoxyacetic acid, unii-wwy23322ik, acetic acid, 4-methylphenoxy, 2-p-tolyloxy acetic acid, p-toloxyacetic acid, acmc-20apmz, p-tolyloxy-acetic acid
70329
2-(4-methylphenoxy)acetic acid
CC1=CC=C(C=C1)OCC(=O)O
CAS Min % 98.5
CAS Max % 100.0
Melting Point 140°C to 142°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3(C6H4)OCH2CO2H
Quantity 5 g
Formula Weight 166.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material (4-Methylphenoxy)acetic acid
Show More Show Less
compliance-icons
Product Identifier
  • (4-Methylphenoxy)acetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'