missing translation for 'onlineSavingsMsg'
Learn More

(4-Methylphenoxy)acetic acid, 99%, ACROS Organics™

Brand:  Acros Organics 378270250

Code : 29

Additional Details : CAS Number : 940-64-7 Weight : 0.07500kg


Quantity 25g
Packaging Glass Bottle

 View more versions of this product

Product Code. 10787913

  • £26.10 / 25g


Estimated Shipment
16-03-2018
Add to basket


Specifications

Chemical Identifiers

(4-Methylphenoxy)acetic acid
98.5
≥98.5%
C9H10O3
166.18
940-64-7
100.0
99%
CH3(C6H4)OCH2CO2H
MFCD00014365

Specifications

(4-Methylphenoxy)acetic acid
136° to 139°C
940-64-7
100.0
Glass Bottle
≥98.5%
C9H10O3
166.18
Slightly beige
25g
98.5
Authentic
Crystalline Powder
99%
CH3(C6H4)OCH2CO2H
MFCD00014365
Videos
SDS