missing translation for 'onlineSavingsMsg'
Learn More

4-Methylnicotinic acid hydrochloride, 95%, Thermo Scientific™

Catalog Number p-2702012
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
94015-05-1
C7H6NO2
136.13
MFCD00832700
ZKUZSTXNVMIDCY-UHFFFAOYSA-M
4-methylnicotinic acid hydrochloride, 4-methylpyridine-3-carboxylic acid hydrochloride, 4-methylnicotinic acid hcl, 4-methyl-nicotinic acid hydrochloride, acmc-209wrr, 4-methyl nicotinic acid hydrochloride, 4-methylnicotinic acid hydrochloride;, 4-methylnicotinic acid monohydrochloride, 4-methylnicotinic acid hydrochloride salt, 4-methylnicotinic acid monohydrochloride;
2796506
CC1=CC=NC=C1C([O-])=O
This item is not returnable. View return policy
94015-05-1
C7H6NO2
136.13
MFCD00832700
ZKUZSTXNVMIDCY-UHFFFAOYSA-M
4-methylnicotinic acid hydrochloride, 4-methylpyridine-3-carboxylic acid hydrochloride, 4-methylnicotinic acid hcl, 4-methyl-nicotinic acid hydrochloride, acmc-209wrr, 4-methyl nicotinic acid hydrochloride, 4-methylnicotinic acid hydrochloride;, 4-methylnicotinic acid monohydrochloride, 4-methylnicotinic acid hydrochloride salt, 4-methylnicotinic acid monohydrochloride;
2796506
CC1=CC=NC=C1C([O-])=O
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 4-methylpyridine-3-carboxylic acid;hydrochloride
Formula Weight 173.6
Percent Purity 95%
Chemical Name or Material 4-Methylnicotinic acid hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'