Learn More
4-Methylmorpholine N-oxide, 50 wt.% solution in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 348271000
Code : 29
Additional Details : CAS Number : 7529-22-8 Weight : 0.10000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
4-Methylmorpholine N-oxide | |
7529-22-8,7732-18-5 | |
50.0 | |
Colorless to Yellow | |
118.0°C to 119.0°C | |
Glass bottle | |
MFCD00005947 | |
07,244 | |
15, 6171 | |
Solubility in water: miscible | |
C[N+]1([O-])CCOCC1 | |
117.15 | |
CHEBI:52093 | |
Liquid |
50 wt.% solution in water | |
48.0 | |
-20.0°C | |
1.1300g/mL | |
>100°C | |
C5H11NO2 | |
100 g | |
1.13 | |
4-methylmorpholine n-oxide, 4-methylmorpholine 4-oxide, n-methylmorpholine oxide, n-methylmorpholine n-oxide, morpholine, 4-methyl-, 4-oxide, 4-methylmorpholine-4-oxide, methyl morpholine oxide, n-methylmorpholine-n-oxide, nmo, unii-arc64pkj0f | |
LFTLOKWAGJYHHR-UHFFFAOYSA-N | |
4-methyl-4-oxidomorpholin-4-ium | |
82029 | |
117.15 |
Safety and Handling
- 4-Methylmorpholine N-oxide, 50% wt.% solution in water
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only