missing translation for 'onlineSavingsMsg'
Learn More

4-Methylimidazole, 98%, Thermo Scientific Chemicals

Catalog Number 10460661
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
822-36-6
C4H6N2
82.11
MFCD00005201
XLSZMDLNRCVEIJ-UHFFFAOYSA-N
4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
13195
CHEBI:40035
5-methyl-1H-imidazole
CC1=CN=CN1
This item is not returnable. View return policy
822-36-6
C4H6N2
82.11
MFCD00005201
XLSZMDLNRCVEIJ-UHFFFAOYSA-N
4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
13195
CHEBI:40035
5-methyl-1H-imidazole
CC1=CN=CN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 52.0°C to 56.0°C
Color Brown to Brown-Red
Boiling Point 263.0°C
Flash Point 157°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 50 g
Beilstein 23, 69
Solubility Information Solubility in water: soluble. Other solubilities: soluble in almost all common organic solvents
Formula Weight 82.11
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Methylimidazole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'