missing translation for 'onlineSavingsMsg'
Learn More

4-Methylhomopiperazine-4-aminobenzene trihydrochloride monohydrate, 90%, Thermo Scientific™

Product Code. p-3522536
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10516914

Brand: Thermo Scientific Maybridge CC60364CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 913830-33-8
Molecular Formula C12H24Cl3N3O
Molecular Weight (g/mol) 332.694
MDL Number MFCD09025902
InChI Key RSGOMRQTIZQMIR-UHFFFAOYSA-N
Synonym 4-methylhomopiperazine-4-aminobenzene trihydrochloride monohydrate, 4-4-methyl-1,4-diazepan-1-yl aniline hydrate trihydrochloride, 4-4-methyl-1,4-diazepan-1-yl aniline trihydrochloride hydrate, 1-4-aminophenyl-4-methylhomopiperazine trihydrochloride hydrate, 4-4-methylhomopiperazin-1-yl aniline trihydrochloride monohydrate, 4-4-methyl-1,4-diazepan-1-yl aniline-hydrogen chloride-water 1/3/1
PubChem CID 44119580
IUPAC Name 4-(4-methyl-1,4-diazepan-1-yl)aniline;hydrate;trihydrochloride
SMILES CN1CCCN(CC1)C2=CC=C(C=C2)N.O.Cl.Cl.Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Linear Formula Yes
Quantity 250 mg
Formula Weight 332.71
Percent Purity 90%
Chemical Name or Material 4-Methylhomopiperazine-4-aminobenzene trihydrochloride monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.