missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexanol, 99%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog Number p-161930
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
589-91-3
C7H14O
114.19
MFCD00001449,MFCD00064171,MFCD00064170
MQWCXKGKQLNYQG-UHFFFAOYSA-N
4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
11524
4-methylcyclohexan-1-ol
CC1CCC(O)CC1
This item is not returnable. View return policy
589-91-3
C7H14O
114.19
MFCD00001449,MFCD00064171,MFCD00064170
MQWCXKGKQLNYQG-UHFFFAOYSA-N
4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
11524
4-methylcyclohexan-1-ol
CC1CCC(O)CC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -41.0°C
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 171.0°C to 173.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula CH3C6H10OH
Refractive Index 1.4570 to 1.4590
Quantity 5 g
Beilstein 06, 14
Specific Gravity 0.92
Formula Weight 114.19
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material 4-Methylcyclohexanol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylcyclohexanol, mixture of cis and trans
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'