missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzylamine, 98%, Thermo Scientific Chemicals

Catalog Number p-159718
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
104-84-7
C8H11N
121.18
MFCD00008123
HMTSWYPNXFHGEP-UHFFFAOYSA-N
4-methylbenzylamine, benzenemethanamine, 4-methyl, p-methylbenzylamine, p-tolylmethanamine, 4-methylphenyl methanamine, benzylamine, p-methyl, p-xylylamine, 4-methybenzylamine, 1-4-methylphenyl methanamine, 4-methyl-benzylamine
66035
(4-methylphenyl)methanamine
CC1=CC=C(C=C1)CN
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
104-84-7
C8H11N
121.18
MFCD00008123
HMTSWYPNXFHGEP-UHFFFAOYSA-N
4-methylbenzylamine, benzenemethanamine, 4-methyl, p-methylbenzylamine, p-tolylmethanamine, 4-methylphenyl methanamine, benzylamine, p-methyl, p-xylylamine, 4-methybenzylamine, 1-4-methylphenyl methanamine, 4-methyl-benzylamine
66035
(4-methylphenyl)methanamine
CC1=CC=C(C=C1)CN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 13°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 195°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2NH2
Refractive Index 1.5330 to 1.536
Quantity 5 g
Beilstein 12, 1141
Specific Gravity 0.95
Solubility Information Solubility in water: slightly soluble
Formula Weight 121.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methylbenzylamine, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'