Learn More
Alfa Aesar™ (4-Methylbenzyl)triphenylphosphonium chloride, 98+%
Brand: Alfa Aesar™ A12447.30
Code : NEW
Additional Details : CAS Number : 1530-37-6 Weight : 0.25000kg
Quantity | 250g |
---|
Chemical Identifiers
(4-Methylbenzyl)triphenylphosphonium chloride | |
C26H24ClP | |
MFCD00031581 |
1530-37-6 | |
402.91 |
Specifications
(4-Methylbenzyl)triphenylphosphonium chloride | |
≥98% | |
402.91 | |
260° to 262°C |
1530-37-6 | |
C26H24ClP | |
MFCD00031581 | |
250g |