missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methylbenzyl mercaptan, 97%

Brand:  Alfa Aesar™ A10467.22

Code : NEW

Additional Details : CAS Number : 4498-99-1 Weight : 0.10000kg Transport : UN number : 2810 Chem class : 1 Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 11438703

  • £445.70 / 100g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Description

Description

Applications
4-Methylbenzyl mercaptan is used as pharmaceutical intermediates.

Solubility
Sparingly Soluble.

Notes
Air Sensitive. Store away from air and oxidizing agents.
Specifications

Chemical Identifiers

4-Methylbenzyl mercaptan
C8H10S
MFCD00039652
2411317
4498-99-1
138.23
UN2810
4-Methyl-alpha-toluenethiol

Specifications

4-Methylbenzyl mercaptan
97%
138.23
UN2810
4-Methyl-alpha-toluenethiol
100g
4498-99-1
C8H10S
MFCD00039652
2411317
116° to 118°C (35mmHg)
Stench
Videos
SDS