missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl cyanide, 98%, Thermo Scientific Chemicals

Catalog Number p-3758342
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2947-61-7
C9H9N
131.18
MFCD00001922
RNHKXHKUKJXLAU-UHFFFAOYSA-N
4-methylbenzyl cyanide, 4-methylphenylacetonitrile, p-tolylacetonitrile, p-methylbenzyl cyanide, 2-4-methylphenyl acetonitrile, benzeneacetonitrile, 4-methyl, 4-methylphenyl acetonitrile, 4-methylbenzeneacetonitrile, 4-methylbenzylcyanide, 2-p-tolylacetonitrile
76280
2-(4-methylphenyl)acetonitrile
CC1=CC=C(CC#N)C=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2947-61-7
C9H9N
131.18
MFCD00001922
RNHKXHKUKJXLAU-UHFFFAOYSA-N
4-methylbenzyl cyanide, 4-methylphenylacetonitrile, p-tolylacetonitrile, p-methylbenzyl cyanide, 2-4-methylphenyl acetonitrile, benzeneacetonitrile, 4-methyl, 4-methylphenyl acetonitrile, 4-methylbenzeneacetonitrile, 4-methylbenzylcyanide, 2-p-tolylacetonitrile
76280
2-(4-methylphenyl)acetonitrile
CC1=CC=C(CC#N)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 18°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 242°C to 243°C
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2CN
Refractive Index 1.5170 to 1.5195
Quantity 5 g
Beilstein 09, 530
Specific Gravity 0.99
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether,benzene
Formula Weight 131.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methylbenzyl cyanide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylbenzyl cyanide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'