missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number 11413407
Click to view available options
Quantity:
25 g
50 g
250 g
This item is not returnable. View return policy
104-82-5
C8H9Cl
140.61
MFCD00000919
DMHZDOTYAVHSEH-UHFFFAOYSA-N
4-methylbenzyl chloride, p-xylyl chloride, 1-chloromethyl-4-methylbenzene, alpha-chloro-p-xylene, p-methylbenzyl chloride, benzene, 1-chloromethyl-4-methyl, p-chloromethyl toluene, 4-chloromethyl toluene, p-tolylmethyl chloride, p-xylene, alpha-chloro
7722
1-(chloromethyl)-4-methylbenzene
CC1=CC=C(C=C1)CCl
This item is not returnable. View return policy
104-82-5
C8H9Cl
140.61
MFCD00000919
DMHZDOTYAVHSEH-UHFFFAOYSA-N
4-methylbenzyl chloride, p-xylyl chloride, 1-chloromethyl-4-methylbenzene, alpha-chloro-p-xylene, p-methylbenzyl chloride, benzene, 1-chloromethyl-4-methyl, p-chloromethyl toluene, 4-chloromethyl toluene, p-tolylmethyl chloride, p-xylene, alpha-chloro
7722
1-(chloromethyl)-4-methylbenzene
CC1=CC=C(C=C1)CCl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 4°C to 6°C
Boiling Point 199°C to 201°C
Flash Point 75°C (167°F)
Odor Pungent
Quantity 50 g
UN Number UN3265
Beilstein 507387
Merck Index 14,10091
Formula Weight 140.61
Percent Purity 98%
Chemical Name or Material 4-Methylbenzyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'