missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl bromide, 98%, Thermo Scientific Chemicals

Catalog Number p-7021101
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
104-81-4
C8H9Br
185.064
MFCD00000181
WZRKSPFYXUXINF-UHFFFAOYSA-N
4-methylbenzyl bromide, 1-bromomethyl-4-methylbenzene, p-xylyl bromide, p-methylbenzyl bromide, alpha-bromo-p-xylene, benzene, 1-bromomethyl-4-methyl, 4-bromomethyl toluene, p-bromomethyl toluene, alpha-bromo-p-xylol, 1-bromomethyl-4-methyl-benzene
7721
1-(bromomethyl)-4-methylbenzene
CC1=CC=C(C=C1)CBr
This item is not returnable. View return policy
104-81-4
C8H9Br
185.064
MFCD00000181
WZRKSPFYXUXINF-UHFFFAOYSA-N
4-methylbenzyl bromide, 1-bromomethyl-4-methylbenzene, p-xylyl bromide, p-methylbenzyl bromide, alpha-bromo-p-xylene, benzene, 1-bromomethyl-4-methyl, 4-bromomethyl toluene, p-bromomethyl toluene, alpha-bromo-p-xylol, 1-bromomethyl-4-methyl-benzene
7721
1-(bromomethyl)-4-methylbenzene
CC1=CC=C(C=C1)CBr

4-Methylbenzyl bromide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbenzyl bromide is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
TRUSTED_SUSTAINABILITY
Melting Point 33°C to 36°C
Density 1.324
Boiling Point 218°C to 220°C
Flash Point 97°C (206°F)
Quantity 5 g
UN Number UN3417
Beilstein 507389
Merck Index 14,10090
Solubility Information Insoluble in water.
Formula Weight 185.07
Percent Purity 98%
Chemical Name or Material 4-Methylbenzyl bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'