missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog Number 10486800
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
589-18-4
C8H10O
122.17
MFCD00004664
KMTDMTZBNYGUNX-UHFFFAOYSA-N
4-methylbenzyl alcohol, p-tolylmethanol, p-tolylcarbinol, 4-methylphenyl methanol, 4-tolylcarbinol, p-methylbenzyl alcohol, benzenemethanol, 4-methyl, 4-hydroxymethyl toluene, benzyl alcohol, p-methyl, 4-methylbenzenemethanol
11505
CHEBI:1895
(4-methylphenyl)methanol
CC1=CC=C(C=C1)CO
This item is not returnable. View return policy
589-18-4
C8H10O
122.17
MFCD00004664
KMTDMTZBNYGUNX-UHFFFAOYSA-N
4-methylbenzyl alcohol, p-tolylmethanol, p-tolylcarbinol, 4-methylphenyl methanol, 4-tolylcarbinol, p-methylbenzyl alcohol, benzenemethanol, 4-methyl, 4-hydroxymethyl toluene, benzyl alcohol, p-methyl, 4-methylbenzenemethanol
11505
CHEBI:1895
(4-methylphenyl)methanol
CC1=CC=C(C=C1)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 56.0°C to 61.0°C
Color White
Boiling Point 217.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2OH
Quantity 100 g
Beilstein 06, 498
Formula Weight 122.17
Percent Purity 98%
Physical Form Crystalline Mass, Crystals or Needles
Chemical Name or Material 4-Methylbenzyl alcohol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylbenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'