missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl alcohol, 98%, Acros Organics

£13.00 - £56.50

Chemical Identifiers

CAS 589-18-4
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00004664
InChI Key KMTDMTZBNYGUNX-UHFFFAOYSA-N
Synonym 4-methylbenzyl alcohol, p-tolylmethanol, p-tolylcarbinol, 4-methylphenyl methanol, 4-tolylcarbinol, p-methylbenzyl alcohol, benzenemethanol, 4-methyl, 4-hydroxymethyl toluene, benzyl alcohol, p-methyl, 4-methylbenzenemethanol
PubChem CID 11505
ChEBI CHEBI:1895
IUPAC Name (4-methylphenyl)methanol
SMILES CC1=CC=C(C=C1)CO
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10035973 SDS View Documents Acros Organics
155920050
5g Glass bottle £13.00
5g View alternative products
Save Up to 
»
EU Stock 4 Please to purchase this item. Need a web account? Register with us today!
10677122 SDS View Documents Acros Organics
155920250
25g Glass bottle £19.20
25g View alternative products
Save Up to 
»
EU Stock 3 Please to purchase this item. Need a web account? Register with us today!
10486800 SDS View Documents Acros Organics
155921000
100g Glass bottle £56.50
100g View alternative products
Save Up to 
»
EU Stock 1 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

589-18-4
122.17
KMTDMTZBNYGUNX-UHFFFAOYSA-N
11505
(4-methylphenyl)methanol
C8H10O
MFCD00004664
4-methylbenzyl alcohol, p-tolylmethanol, p-tolylcarbinol, 4-methylphenyl methanol, 4-tolylcarbinol, p-methylbenzyl alcohol, benzenemethanol, 4-methyl, 4-hydroxymethyl toluene, benzyl alcohol, p-methyl, 4-methylbenzenemethanol
CHEBI:1895
CC1=CC=C(C=C1)CO

Specifications

4-Methylbenzyl alcohol
Glass bottle
White
5g
589-18-4
100.0
C8H10O
MFCD00004664
4-methylbenzyl alcohol, p-tolylmethanol, p-tolylcarbinol, 4-methylphenyl methanol, 4-tolylcarbinol, p-methylbenzyl alcohol, benzenemethanol, 4-methyl, 4-hydroxymethyl toluene, benzyl alcohol, p-methyl, 4-methylbenzenemethanol
CC1=CC=C(C=C1)CO
122.17
CHEBI:1895
Crystalline Mass, Needles, or Crystals
Authentic
217.0°C
56.0°C to 61.0°C
98%
97.5
97.5% min. (GC)
CH3C6H4CH2OH
06, 498
KMTDMTZBNYGUNX-UHFFFAOYSA-N
(4-methylphenyl)methanol
11505
122.17
98%
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Methylbenzyl alcohol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.