missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylbenzophenone, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
500 g
Description
4-Methylbenzophenone is used as clear coatings for wood, plastic and metal, overprint varnish.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methylbenzophenone is used as clear coatings for wood, plastic and metal, overprint varnish.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place. Store in a tightly closed container. Incompatible with strong oxidizing agents.
4-Methylbenzophenone is used as clear coatings for wood, plastic and metal, overprint varnish.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place. Store in a tightly closed container. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 53°C to 57°C |
Boiling Point | 326°C |
Flash Point | 143°C (289°F) |
Quantity | 50 g |
Beilstein | 1909310 |
Merck Index | 14,7317 |
Solubility Information | Insoluble in water. |
Formula Weight | 196.25 |
Percent Purity | 98% |
Chemical Name or Material | 4-Methylbenzophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'