missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methylbenzenethiol sodium salt, 98%

Brand:  Alfa Aesar™ H55095.14

Code : NEW

Additional Details : CAS Number : 10486-08-5 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15481238

  • £152.00 / 25g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Description

Description

Applications
4-Methylbenzenethiol sodium salt is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

4-Methylbenzenethiol sodium salt
10486-08-5
146.18
MFCD00066208

Specifications

4-Methylbenzenethiol sodium salt
>300°C
10486-08-5
98%
MFCD00066208
White to off-white
25g
Powder
146.18
Videos
SDS