Learn More
Alfa Aesar™ 4-Methylbenzenethiol sodium salt, 98%
Brand: Alfa Aesar™ H55095.06
Code : NEW
Additional Details : CAS Number : 10486-08-5 Weight : 0.00500kg
Quantity | 5g |
---|
Description
Applications4-Methylbenzenethiol sodium salt is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
4-Methylbenzenethiol sodium salt | |
10486-08-5 | |
146.18 | |
MFCD00066208 |
Specifications
4-Methylbenzenethiol sodium salt | |
>300°C | |
10486-08-5 | |
98% | |
MFCD00066208 |
White to off-white | |
5g | |
Powder | |
146.18 |