missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Methylaminomethyl)benzonitrile, 95%

Brand:  Alfa Aesar™ H63890.03

Code : NEW

Additional Details : CAS Number : 34403-48-0 Weight : 0.00100kg Transport : UN number : 3276 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15490839

  • £59.78 / 1g


Estimated Shipment date
from Supplier 22-05-2018
Add to basket

Specifications

Chemical Identifiers

4-(Methylaminomethyl)benzonitrile
164.19
UN3276
34403-48-0
MFCD09738490

Specifications

4-(Methylaminomethyl)benzonitrile
34403-48-0
95%
MFCD09738490
1g
Oil
164.19
UN3276
Videos
SDS