missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)phenol sulfate, ACS, 99.0-101.5%, Thermo Scientific Chemicals

Catalog Number p-7050387
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
55-55-0
MFCD00013140
Bis(4-hydroxy-n-methylanilinium) sulfate; Metol
This item is not returnable. View return policy
55-55-0
MFCD00013140
Bis(4-hydroxy-n-methylanilinium) sulfate; Metol

4-(Methylamino)phenol sulfate is a photographic developer used in black and white photography. It is also used as tone developers in developing agents. Further, it is used in oxidative hair dye formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼260°C (decomposition)
Color Cream to White
Quantity 50 g
UN Number UN2811
Beilstein 3919382
Sensitivity Air and light sensitive
Merck Index 14,6017
Solubility Information Soluble in alcohol and ether. Slightly soluble in water and ethanol. Insoluble in diethyl ether.
Formula Weight 344.38
Percent Purity 99.0 to 101.5%
Grade ACS
Physical Form Powder
Chemical Name or Material 4-(Methylamino)phenol sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'