missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7082138
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
10541-83-0
C8H9NO2
151.165
MFCD00002535
ZVIDMSBTYRSMAR-UHFFFAOYSA-N
4-methylamino benzoic acid, benzoic acid, 4-methylamino, p-methylaminobenzoic acid, n-methyl-4-aminobenzoic acid, 4-n-methylamino benzoic acid, n-methyl-4-aminobenzoate, benzoic acid, p-methylamino, 4-methylamino-benzoic acid, p-methylamino benzoic acid, p-n-methylamino benzoic acid
66345
CHEBI:7308
4-(methylamino)benzoic acid
CNC1=CC=C(C=C1)C(=O)O
This item is not returnable. View return policy
10541-83-0
C8H9NO2
151.165
MFCD00002535
ZVIDMSBTYRSMAR-UHFFFAOYSA-N
4-methylamino benzoic acid, benzoic acid, 4-methylamino, p-methylaminobenzoic acid, n-methyl-4-aminobenzoic acid, 4-n-methylamino benzoic acid, n-methyl-4-aminobenzoate, benzoic acid, p-methylamino, 4-methylamino-benzoic acid, p-methylamino benzoic acid, p-n-methylamino benzoic acid
66345
CHEBI:7308
4-(methylamino)benzoic acid
CNC1=CC=C(C=C1)C(=O)O

4-(Methylamino)benzoic acid important for the preparation of other pharmaceutical products, dyes, flavors, and preservatives. .For the preparation of medicines and other fine chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylamino)benzoic acid important for the preparation of other pharmaceutical products, dyes, flavors, and preservatives. .For the preparation of medicines and other fine chemicals.

Solubility
Slightly soluble. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 158°C to 160°C
Quantity 5 g
Beilstein 2082805
Solubility Information Slightly soluble. Soluble in alcohol.
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material 4- (Methylamino)benzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'