missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-4H-thieno[3,2-b]pyrrol-5-yl)methanol, 97%, Thermo Scientific™

Catalog Number p-3109910
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
121933-59-3
C8H9NOS
167.23
MFCD07772838
ZRWPUHUHJRQLCD-UHFFFAOYSA-N
4-methyl-4h-thieno 3,2-b pyrrol-5-yl methanol, 4-methylthieno 3,2-b pyrrol-5-yl methanol, 4h-thieno 3,2-b pyrrole-5-methanol,4-methyl, acmc-1c2v7, 4-methyl-4h-thieno 3,2-b pyrrol-5-yl-methanol, 4-methylthiopheno 2,3-d pyrrol-5-yl methan-1-ol
6483918
{4-methyl-4H-thieno[3,2-b]pyrrol-5-yl}methanol
CN1C(CO)=CC2=C1C=CS2
This item is not returnable. View return policy
121933-59-3
C8H9NOS
167.23
MFCD07772838
ZRWPUHUHJRQLCD-UHFFFAOYSA-N
4-methyl-4h-thieno 3,2-b pyrrol-5-yl methanol, 4-methylthieno 3,2-b pyrrol-5-yl methanol, 4h-thieno 3,2-b pyrrole-5-methanol,4-methyl, acmc-1c2v7, 4-methyl-4h-thieno 3,2-b pyrrol-5-yl-methanol, 4-methylthiopheno 2,3-d pyrrol-5-yl methan-1-ol
6483918
{4-methyl-4H-thieno[3,2-b]pyrrol-5-yl}methanol
CN1C(CO)=CC2=C1C=CS2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 167.23
Percent Purity 97%
Chemical Name or Material (4-Methyl-4H-thieno[3, 2-b]pyrrol-5-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'