missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methylamine, 90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 179.22 |
Percent Purity | 90% |
Chemical Name or Material | (4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'