Learn More
(4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methanol, 97%, Maybridge™
Brand: Maybridge CC62009CB
Code : 31
Additional Details : CAS Number : 921938-81-0 Weight : 0.05025kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 250mg |
Chemical Identifiers
921938-81-0 | |
180.207 | |
PPBFQXGUORHMBA-UHFFFAOYSA-N | |
24229648 | |
CN1CCOC2=C1N=CC(=C2)CO |
C9H12N2O2 | |
MFCD09817507 | |
4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methan-1-ol, 2h-pyrido 3,2-b-1,4-oxazine-7-methanol,3,4-dihydro-4-methyl | |
(4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanol |
Specifications
(4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methanol | |
250mg | |
96.5 | |
97% | |
MFCD09817507 | |
PPBFQXGUORHMBA-UHFFFAOYSA-N | |
(4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanol | |
24229648 | |
97% |
Amber Glass Bottle | |
921938-81-0 | |
100.0 | |
C9H12N2O2 | |
4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methan-1-ol, 2h-pyrido 3,2-b-1,4-oxazine-7-methanol,3,4-dihydro-4-methyl | |
CN1CCOC2=C1N=CC(=C2)CO | |
180.207 | |
180.21 |