missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-6-sulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-3629705
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
892948-94-6
C9H10ClNO3S
247.69
MFCD10700049
LHMOALYDXXCXLZ-UHFFFAOYSA-N
4-methyl-3,4-dihydro-2h-1,4-benzoxazine-6-sulfonyl chloride, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-6-sulfonyl chloride, 2h-1,4-benzoxazine-6-sulfonyl chloride, 3,4-dihydro-4-methyl, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-6-sulfonylchloride, 4-methyl-3,4-dihydro-2h-1,4-benzoxazine-6-sulfonyl, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-6-sulfonyl chloride, 4-methyl-3,4-dihydro-2h-benzo 1,4 oxazine-6-sulfonyl chloride, chloro 4-methyl 2h,3h-benzo 3,4-e 1,4-oxazaperhydroin-6-yl sulfone
43811036
4-methyl-2,3-dihydro-1,4-benzoxazine-6-sulfonyl chloride
CN1CCOC2=C1C=C(C=C2)S(Cl)(=O)=O
This item is not returnable. View return policy
892948-94-6
C9H10ClNO3S
247.69
MFCD10700049
LHMOALYDXXCXLZ-UHFFFAOYSA-N
4-methyl-3,4-dihydro-2h-1,4-benzoxazine-6-sulfonyl chloride, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-6-sulfonyl chloride, 2h-1,4-benzoxazine-6-sulfonyl chloride, 3,4-dihydro-4-methyl, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-6-sulfonylchloride, 4-methyl-3,4-dihydro-2h-1,4-benzoxazine-6-sulfonyl, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-6-sulfonyl chloride, 4-methyl-3,4-dihydro-2h-benzo 1,4 oxazine-6-sulfonyl chloride, chloro 4-methyl 2h,3h-benzo 3,4-e 1,4-oxazaperhydroin-6-yl sulfone
43811036
4-methyl-2,3-dihydro-1,4-benzoxazine-6-sulfonyl chloride
CN1CCOC2=C1C=C(C=C2)S(Cl)(=O)=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 247.7
Percent Purity 97%
Chemical Name or Material 4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-6-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'