missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-3,4-dihydro-2H-1,4-benzoxazin-7-yl)methylamine, 97%, Thermo Scientific™

Catalog Number p-3629683
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
946409-08-1
C10H14N2O
178.235
MFCD11109315
PGIOCCIKSFJJMR-UHFFFAOYSA-N
4-methyl-3,4-dihydro-2h-1,4-benzoxazin-7-yl methylamine, 4-methyl-2,3-dihydro-1,4-benzoxazin-7-yl methanamine, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazin-7-yl methanamine, 1-4-methyl-2,3-dihydro-1,4-benzoxazin-7-yl methanamine, 4-methyl-2h,3h-benzo e 1,4-oxazin-7-yl methylamine, 1-4-methyl-3,4-dihydro-2h-1,4-benzoxazin-7-yl methanamine
33589445
(4-methyl-2,3-dihydro-1,4-benzoxazin-7-yl)methanamine
CN1CCOC2=C1C=CC(=C2)CN
This item is not returnable. View return policy
946409-08-1
C10H14N2O
178.235
MFCD11109315
PGIOCCIKSFJJMR-UHFFFAOYSA-N
4-methyl-3,4-dihydro-2h-1,4-benzoxazin-7-yl methylamine, 4-methyl-2,3-dihydro-1,4-benzoxazin-7-yl methanamine, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazin-7-yl methanamine, 1-4-methyl-2,3-dihydro-1,4-benzoxazin-7-yl methanamine, 4-methyl-2h,3h-benzo e 1,4-oxazin-7-yl methylamine, 1-4-methyl-3,4-dihydro-2h-1,4-benzoxazin-7-yl methanamine
33589445
(4-methyl-2,3-dihydro-1,4-benzoxazin-7-yl)methanamine
CN1CCOC2=C1C=CC(=C2)CN
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material (4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazin-7-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'